siRNA Details
| Gene Symbol: Gm9767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028030.2 | 1 | 1048 | |||
| GenBank mRNA | AK021041.1 | - | 1 | 1049 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm9767| Gene Symbol: Gm9767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028030.2 | 1 | 1048 | |||
| GenBank mRNA | AK021041.1 | - | 1 | 1049 | ||