siRNA Details
| Gene Symbol: 4930567H12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015535.2 | 3 | 942 | |||
| GenBank mRNA | AK131761.1 | - | 3 | 943 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930567H12Rik| Gene Symbol: 4930567H12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015535.2 | 3 | 942 | |||
| GenBank mRNA | AK131761.1 | - | 3 | 943 | ||