siRNA Details
| Gene Symbol: 4930583K01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027879.1 | 2 | 861 | |||
| GenBank mRNA | BB019113.1 | - | 1 | 212 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930583K01Rik| Gene Symbol: 4930583K01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027879.1 | 2 | 861 | |||
| GenBank mRNA | BB019113.1 | - | 1 | 212 | ||