siRNA Details
Gene Symbol: Mgarp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028121.1 | 1 | 297 | |||
GenBank mRNA | BC022740.1 | - | 1 | 252 | 4511440 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mgarp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028121.1 | 1 | 297 | |||
GenBank mRNA | BC022740.1 | - | 1 | 252 | 4511440 |