siRNA Details
| Gene Symbol: 4930584F24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029445.1 | 2 | 208 | |||
| GenBank mRNA | AK016349.1 | - | 2 | 175 | ||
| BC049642.1 | - | 2 | 183 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU935604.1 | - | 2 | 186 | Chr. 5 - 26479718 - 26493289 [-] on Build GRCm38 | |

4930584F24Rik
Genomic Map