siRNA Details
| Gene Symbol: 4930584F24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029445.1 | 3 | 582 | |||
| GenBank mRNA | AK005600.1 | - | 4 | 515 | ||
| AK015867.1 | - | 2 | 280 | |||
| BC049642.1 | - | 3 | 557 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930584F24Rik| Gene Symbol: 4930584F24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029445.1 | 3 | 582 | |||
| GenBank mRNA | AK005600.1 | - | 4 | 515 | ||
| AK015867.1 | - | 2 | 280 | |||
| BC049642.1 | - | 3 | 557 | |||