siRNA Details
| Gene Symbol: Cts8-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027871.2 | 5 | 3780 | |||
| GenBank mRNA | BC050022.1 | - | 5 | 3782 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cts8-ps| Gene Symbol: Cts8-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027871.2 | 5 | 3780 | |||
| GenBank mRNA | BC050022.1 | - | 5 | 3782 | ||