siRNA Details
| Gene Symbol: Gm17762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028378.1 | 1 | 2067 | |||
| GenBank mRNA | AK033161.1 | - | 1 | 2068 | ||
| AK045818.1 | - | 1 | 386 | |||
| AK171729.1 | - | 1 | 1758 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm17762| Gene Symbol: Gm17762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028378.1 | 1 | 2067 | |||
| GenBank mRNA | AK033161.1 | - | 1 | 2068 | ||
| AK045818.1 | - | 1 | 386 | |||
| AK171729.1 | - | 1 | 1758 | |||