siRNA Details
| Gene Symbol: Mir17hg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029382.1 | 1 | 1464 | |||
| GenBank mRNA | AK017164.1 | - | 1 | 328 | ||
| AK053349.1 | - | 1 | 1357 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mir17hg| Gene Symbol: Mir17hg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029382.1 | 1 | 1464 | |||
| GenBank mRNA | AK017164.1 | - | 1 | 328 | ||
| AK053349.1 | - | 1 | 1357 | |||