siRNA Details
| Gene Symbol: Bach2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026843.1 | 5 | 1393 | |||
| GenBank mRNA | BC024582.1 | - | 6 | 1365 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bach2os| Gene Symbol: Bach2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026843.1 | 5 | 1393 | |||
| GenBank mRNA | BC024582.1 | - | 6 | 1365 | ||