siRNA Details
| Gene Symbol: Zfp57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033137.1 | 1 | 156 | |||
| GenBank mRNA | AK143118.1 | - | 1 | 156 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp57| Gene Symbol: Zfp57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033137.1 | 1 | 156 | |||
| GenBank mRNA | AK143118.1 | - | 1 | 156 | ||