siRNA Details
| Gene Symbol: Emx2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_002863.2 | 4 | 4076 | |||
| GenBank mRNA | AY117414.1 | - | LIT1111 EMX2OS |
4 | 4087 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Emx2os| Gene Symbol: Emx2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_002863.2 | 4 | 4076 | |||
| GenBank mRNA | AY117414.1 | - | LIT1111 EMX2OS |
4 | 4087 | |