siRNA Details
| Gene Symbol: Pex16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028114.1 | 1 | 48 | |||
| GenBank mRNA | AK020743.1 | - | 1 | 49 | ||
| AK162716.1 | - | 1 | 39 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY724441.1 | - | 1 | 42 | Chr. 2 - 92374682 - 92378848 [+] on Build GRCm38 | |

Pex16
Genomic Map