siRNA Details
| Gene Symbol: 6530411M01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027881.1 | 1, 2 | 729 | |||
| GenBank mRNA | AK078247.1 | - | 1, 2 | 544 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY718979.1 | - | 1, 2 | 544 | Chr. 17 - 9149174 - 9167836 [-] on Build GRCm38 | |

6530411M01Rik
Genomic Map