siRNA Details
| Gene Symbol: 6530411M01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027881.1 | 3 | 1195 | |||
| GenBank mRNA | AK078247.1 | - | 3 | 1010 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6530411M01Rik| Gene Symbol: 6530411M01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027881.1 | 3 | 1195 | |||
| GenBank mRNA | AK078247.1 | - | 3 | 1010 | ||