siRNA Details
| Gene Symbol: 1700029M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015613.1 | 3 | 2605 | |||
| NR_027909.1 | 2 | 2476 | ||||
| GenBank mRNA | AK006510.2 | - | 3 | 2490 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700029M20Rik| Gene Symbol: 1700029M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015613.1 | 3 | 2605 | |||
| NR_027909.1 | 2 | 2476 | ||||
| GenBank mRNA | AK006510.2 | - | 3 | 2490 | ||