siRNA Details
| Gene Symbol: Vmn1r-ps79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030707.1 | 1 | 680 | |||
| GenBank mRNA | BC115870.1 | - | 1 | 581 | 40062110 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r-ps79| Gene Symbol: Vmn1r-ps79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030707.1 | 1 | 680 | |||
| GenBank mRNA | BC115870.1 | - | 1 | 581 | 40062110 | |