siRNA Details
| Gene Symbol: Klk1b7-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033120.1 | 3 | 429 | |||
| GenBank mRNA | AK007298.1 | - | 431 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klk1b7-ps| Gene Symbol: Klk1b7-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033120.1 | 3 | 429 | |||
| GenBank mRNA | AK007298.1 | - | 431 | |||