siRNA Details
| Gene Symbol: Speer9-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001583.3 | 4 | 751 | |||
| GenBank mRNA | BY714690.1 | - | 4 | 752 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK015142.1 | - | 753 | Chr. - - [-] on Build | ||

Speer9-ps1
Genomic Map