siRNA Details
| Gene Symbol: E330033B04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030690.1 | 1 | 1514 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E330033B04Rik| Gene Symbol: E330033B04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030690.1 | 1 | 1514 | |||