siRNA Details
Gene Symbol: 6330549D23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003619.2 | 2 | 643 | |||
GenBank mRNA | AK032024.1 | - | 2 | 643 | ||
AK088233.1 | - | 3 | 1043 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 6330549D23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003619.2 | 2 | 643 | |||
GenBank mRNA | AK032024.1 | - | 2 | 643 | ||
AK088233.1 | - | 3 | 1043 |