siRNA Details
| Gene Symbol: 6330549D23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003619.2 | 2 | 1266 | |||
| GenBank mRNA | AK032024.1 | - | 2 | 1266 | ||
| AK088233.1 | - | 3 | 1666 | |||
| BC147330.1 | - | 1 | 89 | |||
| BC147331.1 | - | 1 | 89 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6330549D23Rik| Gene Symbol: 6330549D23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003619.2 | 2 | 1266 | |||
| GenBank mRNA | AK032024.1 | - | 2 | 1266 | ||
| AK088233.1 | - | 3 | 1666 | |||
| BC147330.1 | - | 1 | 89 | |||
| BC147331.1 | - | 1 | 89 | |||