siRNA Details
| Gene Symbol: LOC106740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027905.1 | 1 | 21 | |||
| GenBank mRNA | AK134370.1 | - | 1 | 21 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR759674.1 | - | 1 | 188 | Chr. 17 - 14948075 - 14948990 [-] on Build GRCm38 | |

LOC106740
Genomic Map