siRNA Details
| Gene Symbol: LOC106740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027905.1 | 1 | 789 | |||
| GenBank mRNA | AK134370.1 | - | 1 | 789 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV024517.2 | - | 1 | 614 | Chr. 17 - 14947738 - 14948648 [-] on Build GRCm38 | |

LOC106740
Genomic Map