siRNA Details
| Gene Symbol: Gm11346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024599.1 | 2 | 972 | |||
| GenBank mRNA | AK017957.1 | - | 1 | 746 | ||
| AK139092.1 | - | 2182 | ||||
| BC064072.1 | - | 2 | 972 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ236201.1 | - | 1 | 425 | Chr. 13 - 24598030 - 24598693 [-] on Build GRCm38 | |

Gm11346
Genomic Map