siRNA Details
| Gene Symbol: Gm11346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024599.1 | 2 | 492 | |||
| GenBank mRNA | AK017957.1 | - | 1 | 266 | ||
| AK139092.1 | - | 1702 | ||||
| BC064072.1 | - | 2 | 492 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm11346| Gene Symbol: Gm11346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024599.1 | 2 | 492 | |||
| GenBank mRNA | AK017957.1 | - | 1 | 266 | ||
| AK139092.1 | - | 1702 | ||||
| BC064072.1 | - | 2 | 492 | |||