siRNA Details
| Gene Symbol: Gm11346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024599.1 | 1 | 90 | |||
| GenBank mRNA | AK139092.1 | - | 248 | |||
| BC064072.1 | - | 1 | 90 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA125112.1 | - | 1 | 79 | Chr. 13 - 24598793 - 24604755 [-] on Build GRCm38 | |

Gm11346
Genomic Map