siRNA Details
| Gene Symbol: Kis2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_003188.1 | 1 | 1876 | |||
| GenBank mRNA | AK084356.1 | - | 2 | 1308 | ||
| AY940614.1 | - | LIT3387 Kis2 |
1 | 1876 | ||
| AY940615.1 | - | LIT3388 Kis2 |
2 | 1562 | ||
| AY940616.1 | - | LIT3389 Kis2 |
1 | 1195 | ||

Kis2
Genomic Map