siRNA Details
| Gene Symbol: Snhg8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028574.1 | 1 | 564 | |||
| GenBank mRNA | AK008369.1 | - | 1 | 15 | ||
| BC025067.1 | - | 2 | 311 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snhg8| Gene Symbol: Snhg8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028574.1 | 1 | 564 | |||
| GenBank mRNA | AK008369.1 | - | 1 | 15 | ||
| BC025067.1 | - | 2 | 311 | |||