siRNA Details
| Gene Symbol: 1700052K11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027956.1 | 1 | 1723 | |||
| GenBank mRNA | AK148336.1 | - | 1 | 1598 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700052K11Rik| Gene Symbol: 1700052K11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027956.1 | 1 | 1723 | |||
| GenBank mRNA | AK148336.1 | - | 1 | 1598 | ||