siRNA Details
| Gene Symbol: 1700052K11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027956.1 | 1 | 1287 | |||
| GenBank mRNA | AK006771.1 | - | 1 | 1285 | ||
| AK148336.1 | - | 1 | 1162 | |||
| BC099431.1 | - | 1 | 969 | 30525202 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_028539.1 | - | 1285 | Chr. - - [-] on Build | ||

1700052K11Rik
Genomic Map