siRNA Details
| Gene Symbol: Sprr2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003548.1 | 2 | 387 | |||
| GenBank mRNA | AY158991.1 | - | 2 | 386 | ||
| BC130233.1 | - | 2 | 370 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sprr2g| Gene Symbol: Sprr2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003548.1 | 2 | 387 | |||
| GenBank mRNA | AY158991.1 | - | 2 | 386 | ||
| BC130233.1 | - | 2 | 370 | |||