siRNA Details
| Gene Symbol: Smok4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030763.1 | 2 | 1213 | |||
| GenBank mRNA | AK029728.1 | - | 2 | 1214 | ||
| BC125398.1 | - | 1 | 772 | |||
| BC132092.1 | - | 1 | 772 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smok4a| Gene Symbol: Smok4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030763.1 | 2 | 1213 | |||
| GenBank mRNA | AK029728.1 | - | 2 | 1214 | ||
| BC125398.1 | - | 1 | 772 | |||
| BC132092.1 | - | 1 | 772 | |||