siRNA Details
| Gene Symbol: Sprr2j-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003185.1 | 2 | 549 | |||
| GenBank mRNA | AK019074.1 | - | 2 | 551 | ||
| AY158994.1 | - | 2 | 554 | |||
| BC125542.1 | - | 2 | 525 | |||
| BC125544.1 | - | 2 | 525 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY708877.1 | - | 2 | 551 | Chr. 3 - 92418087 - 92419398 [+] on Build GRCm38 | |

Sprr2j-ps
Genomic Map