siRNA Details
| Gene Symbol: Fcho1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028267.1 | 26 | 2676 | |||
| GenBank mRNA | AK014382.1 | - | 26 | 2661 | ||
| AK088957.1 | - | 25 | 2508 | |||
| AK172927.1 | - | 16 | 4154 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fcho1| Gene Symbol: Fcho1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028267.1 | 26 | 2676 | |||
| GenBank mRNA | AK014382.1 | - | 26 | 2661 | ||
| AK088957.1 | - | 25 | 2508 | |||
| AK172927.1 | - | 16 | 4154 | |||