siRNA Details
| Gene Symbol: 2210015D19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015577.1 | 7 | 1650 | |||
| GenBank mRNA | AK005556.1 | - | 7 | 1650 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2210015D19Rik| Gene Symbol: 2210015D19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015577.1 | 7 | 1650 | |||
| GenBank mRNA | AK005556.1 | - | 7 | 1650 | ||