siRNA Details
| Gene Symbol: 2210015D19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015577.1 | 1 | 79 | |||
| GenBank mRNA | AK005556.1 | - | 1 | 79 | ||
| AK008730.1 | - | 1 | 74 | |||
| AK009198.1 | - | 1 | 101 | |||
| BC052911.1 | - | 1 | 63 | |||
| BC094935.1 | - | 1 | 51 | |||

2210015D19Rik
Genomic Map