siRNA Details
| Gene Symbol: B930059L03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033340.1 | 1 | 1203 | |||
| GenBank mRNA | AK083693.1 | - | 1 | 826 | ||
| Gene Symbol: 9430024F10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BB448693.2 | - | 1 | 557 | ||

B930059L03Rik
Genomic Map