siRNA Details
Gene Symbol: B930059L03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033340.1 | 1 | 646 | |||
GenBank mRNA | AK047421.1 | - | 1 | 647 | ||
AK083693.1 | - | 1 | 261 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: B930059L03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033340.1 | 1 | 646 | |||
GenBank mRNA | AK047421.1 | - | 1 | 647 | ||
AK083693.1 | - | 1 | 261 |