siRNA Details
| Gene Symbol: B930059L03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033340.1 | 1 | 756 | |||
| GenBank mRNA | AK047421.1 | - | 1 | 757 | ||
| AK083693.1 | - | 1 | 371 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B930059L03Rik| Gene Symbol: B930059L03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033340.1 | 1 | 756 | |||
| GenBank mRNA | AK047421.1 | - | 1 | 757 | ||
| AK083693.1 | - | 1 | 371 | |||