siRNA Details
| Gene Symbol: 1500011K16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015476.1 | 2 | 409 | |||
| GenBank mRNA | AK005218.1 | - | 2 | 415 | ||
| AK008527.1 | - | 2 | 410 | |||
| AK027942.1 | - | 2 | 410 | |||
| AK165423.1 | - | 2 | 395 | |||
| BC048556.1 | - | 2 | 404 | |||

1500011K16Rik
Genomic Map