siRNA Details
| Gene Symbol: 1500011K16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015476.1 | 2 | 443 | |||
| GenBank mRNA | AK005218.1 | - | 2 | 449 | ||
| AK027942.1 | - | 2 | 444 | |||
| AK165423.1 | - | 2 | 429 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1500011K16Rik| Gene Symbol: 1500011K16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015476.1 | 2 | 443 | |||
| GenBank mRNA | AK005218.1 | - | 2 | 449 | ||
| AK027942.1 | - | 2 | 444 | |||
| AK165423.1 | - | 2 | 429 | |||