siRNA Details
| Gene Symbol: C030034I22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026848.1 | 1 | 457 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE949423.1 | - | 1 | 477 | Chr. 17 - 69416427 - 69416975 [+] on Build GRCm38 | |

C030034I22Rik
Genomic Map