siRNA Details
| Gene Symbol: C030034I22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026848.1 | 1 | 1281 | |||
| GenBank mRNA | AK021121.1 | - | 1 | 331 | ||
| AK147961.1 | - | 328 | ||||
| AK165105.1 | - | 1 | 268 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C030034I22Rik| Gene Symbol: C030034I22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026848.1 | 1 | 1281 | |||
| GenBank mRNA | AK021121.1 | - | 1 | 331 | ||
| AK147961.1 | - | 328 | ||||
| AK165105.1 | - | 1 | 268 | |||