siRNA Details
| Gene Symbol: D430019H16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052429.1 | - | 2 | 910 | ||
| AK081193.1 | - | 2 | 771 | |||
| BC058677.1 | - | 1 | 225 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D430019H16Rik| Gene Symbol: D430019H16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052429.1 | - | 2 | 910 | ||
| AK081193.1 | - | 2 | 771 | |||
| BC058677.1 | - | 1 | 225 | |||