siRNA Details
| Gene Symbol: Gm9871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027989.1 | 4 | 947 | |||
| GenBank mRNA | AK027905.1 | - | 4 | 947 | ||
| BC125571.1 | - | 3 | 580 | |||
| BC144923.1 | - | 3 | 698 | |||
| BC145947.1 | - | 3 | 580 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm9871| Gene Symbol: Gm9871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027989.1 | 4 | 947 | |||
| GenBank mRNA | AK027905.1 | - | 4 | 947 | ||
| BC125571.1 | - | 3 | 580 | |||
| BC144923.1 | - | 3 | 698 | |||
| BC145947.1 | - | 3 | 580 | |||