siRNA Details
| Gene Symbol: Gm9871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027989.1 | 3 | 460 | |||
| GenBank mRNA | AK027905.1 | - | 3 | 460 | ||
| BC125571.1 | - | 2 | 93 | |||
| BC144923.1 | - | 2 | 93 | |||
| BC145947.1 | - | 2 | 93 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm9871| Gene Symbol: Gm9871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027989.1 | 3 | 460 | |||
| GenBank mRNA | AK027905.1 | - | 3 | 460 | ||
| BC125571.1 | - | 2 | 93 | |||
| BC144923.1 | - | 2 | 93 | |||
| BC145947.1 | - | 2 | 93 | |||