siRNA Details
| Gene Symbol: Gm10272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026831.1 | 1 | 18 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10272| Gene Symbol: Gm10272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026831.1 | 1 | 18 | |||