siRNA Details
| Gene Symbol: A930003O13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027362.1 | 3 | 1864 | |||
| GenBank mRNA | BC030424.1 | - | 3 | 1874 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930003O13Rik| Gene Symbol: A930003O13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027362.1 | 3 | 1864 | |||
| GenBank mRNA | BC030424.1 | - | 3 | 1874 | ||