siRNA Details
| Gene Symbol: Hspa13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027492.1 | 5 | 891 | |||
| GenBank mRNA | AK030553.1 | - | 5 | 864 | ||
| BC094508.1 | - | 5 | 834 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hspa13| Gene Symbol: Hspa13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027492.1 | 5 | 891 | |||
| GenBank mRNA | AK030553.1 | - | 5 | 864 | ||
| BC094508.1 | - | 5 | 834 | |||